missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-tert-butylphenylacetic Acid, TRC

Product Code. 30383290
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383290 100 mg
30370212 10 mg
30381309 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383290 Supplier Toronto Research Chemicals Supplier No. TRCD479020100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 854646-92-7
Molecular Formula C14 H20 O2
Molecular Weight (g/mol) 220.31
Synonym [4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetic Acid, Xylometazoline Hydrochloride Imp. F (EP)
IUPAC Name 2-(4-tert-butyl-2,6-dimethylphenyl)acetic acid
SMILES Cc1cc(cc(C)c1CC(=O)O)C(C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H20O2/c1-9-6-11(14(3,4)5)7-10(2)12(9)8-13(15)16/h6-7H,8H2,1-5H3,(H,15,16)
Formula Weight 220.1463
Chemical Name or Material (4-(1,1-Dimethylethyl)-2,6-dimethylphenyl)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.