CAS RN 854646-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2,6-Dimethyl-4-tert-butylphenylacetic Acid, TRC
CAS: 854646-92-7 Molecular Formula: C14 H20 O2 Molecular Weight (g/mol): 220.31 Synonym: [4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetic Acid,Xylometazoline Hydrochloride Imp. F (EP) IUPAC Name: 2-(4-tert-butyl-2,6-dimethylphenyl)acetic acid SMILES: Cc1cc(cc(C)c1CC(=O)O)C(C)(C)C
2
[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.