missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorothiobenzamide, 97%, Thermo Scientific™

Product Code. p-7029197
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15473507

Brand: Thermo Scientific Alfa Aesar H26636.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918-13-4
Molecular Formula C7H5Cl2NS
Molecular Weight (g/mol) 206.08
MDL Number MFCD00053523
InChI Key KGKGSIUWJCAFPX-UHFFFAOYSA-N
Synonym 2,6-dichlorothiobenzamide, chlorthiamid, chlorthiamide, chlorothiamide, chlorthioamide, chlortiamid, prefix, dcbn, benzenecarbothioamide, 2,6-dichloro, 2,6-dichlorobenzothioamide
PubChem CID 2734819
ChEBI CHEBI:949
SMILES NC(=S)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 149°C to 153°C
Flash Point >100°C (212°F)
Quantity 250 mg
Beilstein 1910353
IUPAC Name 2,6-dichlorobenzenecarbothioamide
Formula Weight 206.09
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorothiobenzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.