CAS RN 1918-13-4
CAS RN 1918-13-4
IUPAC Name:
2,6-dichlorobenzene-1-carbothioamide
Synonyms:
2,6-dichlorothiobenzamide chlorthiamide
Molecular Weight (g/mol):
206.08
Molecular Formula:
C7H5Cl2NS
InChi Key:
KGKGSIUWJCAFPX-UHFFFAOYSA-N
SMILES:
NC(=S)C1=C(Cl)C=CC=C1Cl
1
–
3
of
3
results
1
Chlorthiamid, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
2,6-Dichlorothiobenzamide, 97%, Thermo Scientific™
CAS: 1918-13-4 Molecular Formula: C7H5Cl2NS Molecular Weight (g/mol): 206.08 MDL Number: MFCD00053523 InChI Key: KGKGSIUWJCAFPX-UHFFFAOYSA-N Synonym: 2,6-dichlorothiobenzamide,chlorthiamid,chlorthiamide,chlorothiamide,chlorthioamide,chlortiamid,prefix,dcbn,benzenecarbothioamide, 2,6-dichloro,2,6-dichlorobenzothioamide PubChem CID: 2734819 ChEBI: CHEBI:949 SMILES: NC(=S)C1=C(Cl)C=CC=C1Cl