missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15483217
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483217 1 g 1g
15493217 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483217 Supplier Thermo Scientific Alfa Aesar Supplier No. B20541.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5398-44-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00234147
InChI Key SQSYNRCXIZHKAI-UHFFFAOYSA-N
Synonym 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403
PubChem CID 94830
ChEBI CHEBI:73179
IUPAC Name 2,6-dichloropyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(Cl)=NC(Cl)=C1

Specifications

Melting Point 208°C (decomposition)
Quantity 1 g
Formula Weight 192
Percent Purity 98%
Chemical Name or Material 2,6-Dichloropyridine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.