CAS RN 5398-44-7
IUPAC Name:
2,6-dichloropyridine-4-carboxylic acid
Synonyms:
2,6-dichloropyridine-4-carboxylic acid
Molecular Weight (g/mol):
192
Molecular Formula:
C6H3Cl2NO2
InChi Key:
SQSYNRCXIZHKAI-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(Cl)=NC(Cl)=C1
1
–
1
of
1
results
1
2,6-Dichloropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 5398-44-7 Molecular Formula: C6H3Cl2NO2 Molecular Weight (g/mol): 192.00 MDL Number: MFCD00234147 InChI Key: SQSYNRCXIZHKAI-UHFFFAOYSA-N Synonym: 2,6-dichloroisonicotinic acid,2,6-dichloro-isonicotinic acid,2,6-dichloro-4-pyridinecarboxylic acid,4-pyridinecarboxylic acid, 2,6-dichloro,2,6-dichloroisonicotinicacid,2,6-dichloro-4-pyridinyl carboxylic acid,pyridine-4-carboxylic acid, 2,6-dichloro,2,6-dichlorisonicotins,enamine_005327,zlchem 403 PubChem CID: 94830 ChEBI: CHEBI:73179 IUPAC Name: 2,6-dichloropyridine-4-carboxylic acid SMILES: OC(=O)C1=CC(Cl)=NC(Cl)=C1