missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15493217
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15493217

Brand: Thermo Scientific Alfa Aesar B20541.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5398-44-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00234147
InChI Key SQSYNRCXIZHKAI-UHFFFAOYSA-N
Synonym 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403
PubChem CID 94830
ChEBI CHEBI:73179
IUPAC Name 2,6-dichloropyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(Cl)=NC(Cl)=C1

Specifications

Melting Point 208°C (decomposition)
Quantity 5 g
Formula Weight 192
Percent Purity 98%
Chemical Name or Material 2,6-Dichloropyridine-4-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.