missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropurine-9-Beta-D-riboside, TRC

Product Code. 30386488
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386488 100 mg
30377918 250 mg
30374863 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386488 Supplier Toronto Research Chemicals Supplier No. TRCD435785100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13276-52-3
Molecular Formula C10 H10 Cl2 N4 O4
Molecular Weight (g/mol) 321.12
Synonym Regadenoson Impurity 36
IUPAC Name (2R,3R,4S,5R)-2-(2,6-dichloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(Cl)nc(Cl)nc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H10Cl2N4O4/c11-7-4-8(15-10(12)14-7)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2/t3-,5-,6-,9-/m1/s1
Formula Weight 320.0079
Chemical Name or Material 2,6-Dichloropurine-9-beta-D-riboside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.