CAS RN 13276-52-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dichloropurine-9-Beta-D-riboside, TRC
CAS: 13276-52-3 Molecular Formula: C10 H10 Cl2 N4 O4 Molecular Weight (g/mol): 321.12 Synonym: Regadenoson Impurity 36 IUPAC Name: (2R,3R,4S,5R)-2-(2,6-dichloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(Cl)nc(Cl)nc23