missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloroacetanilide, TRC

Product Code. 30380533
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380533 500 mg 500mg
30376210 1 g 1g
30388735 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380533 Supplier Toronto Research Chemicals Supplier No. TRCD431615500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17700-54-8
Molecular Formula C8H7Cl2NO
Molecular Weight (g/mol) 204.05
Synonym N-(2,6-Dichlorophenyl)acetamide, 2',6'-Dichloroacetanilide, N-Acetyl-2,6-dichloroaniline, NSC 158653, 2',6'-Dichloro-acetanilide
SMILES CC(=O)Nc1c(Cl)cccc1Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H7Cl2NO/c1-5(12)11-8-6(9)3-2-4-7(8)10/h2-4H,1H3,(H,11,12)
Formula Weight 202.9905
Chemical Name or Material 2',6'-Dichloroacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.