CAS RN 17700-54-8
IUPAC Name:
N-(2,6-dichlorophenyl)acetamide
Synonyms:
2,6-dichloroacetanilide
Molecular Weight (g/mol):
204.05
Molecular Formula:
C8H7Cl2NO
InChi Key:
DWVWVSLAIJHBBG-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=C(Cl)C=CC=C1Cl
1
–
1
of
1
results
1
2',6'-Dichloroacetanilide, TRC
CAS: 17700-54-8 Molecular Formula: C8H7Cl2NO Molecular Weight (g/mol): 204.05 Synonym: N-(2,6-Dichlorophenyl)acetamide,2',6'-Dichloroacetanilide,N-Acetyl-2,6-dichloroaniline,NSC 158653,2',6'-Dichloro-acetanilide SMILES: CC(=O)Nc1c(Cl)cccc1Cl