missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-nitroanisole, TRC

Product Code. 30383830
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383830 1 g
30388413 500 mg
30369412 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383830 Supplier Toronto Research Chemicals Supplier No. TRCD4384801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17742-69-7
Molecular Formula C7 H5 Cl2 N O3
Molecular Weight (g/mol) 222.03
Synonym 3,5-Dichloro-4-methoxynitrobenzene, NSC 212118, 1,3-Dichloro-2-methoxy-5-nitrobenzene
IUPAC Name 1,3-dichloro-2-methoxy-5-nitro-benzene
SMILES COc1c(Cl)cc(cc1Cl)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5Cl2NO3/c1-13-7-5(8)2-4(10(11)12)3-6(7)9/h2-3H,1H3
Formula Weight 220.9646
Chemical Name or Material 2,6-Dichloro-4-nitroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.