accessibility menu, dialog, popup

UserName

H3COClClNOO

CAS RN 17742-69-7

IUPAC Name: 1,3-dichloro-2-methoxy-5-nitrobenzene
Synonyms: 1,3-dichloro-2-methoxy-5-nitrobenzene
Molecular Weight (g/mol): 222.02
Molecular Formula: C7H5Cl2NO3
InChi Key: GJYVJKPFYCKNEC-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(C=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)