missing translation for 'onlineSavingsMsg'
Learn More

2,6-di-tert-Butyl-4-(1-hydroxyethyl)phenol, TRC

Product Code. 30382662
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382662 500 mg 500mg
30385261 1 g -
30375608 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382662 Supplier Toronto Research Chemicals Supplier No. TRCD555045500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14681-20-0
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
Synonym 3,5-Bis(1,1-dimethylethyl)-4-hydroxy-alpha-methylbenzenemethanol, 2,6-Di-tert-butyl-4-(1-hydroxyethyl)phenol, 2,6-Di-tert-butyl-4-(alpha-hydroxyethyl)phenol
SMILES CC(O)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H26O2/c1-10(17)11-8-12(15(2,3)4)14(18)13(9-11)16(5,6)7/h8-10,17-18H,1-7H3
Formula Weight 250.1933
Chemical Name or Material 2,6-di-tert-Butyl-4-(1-hydroxyethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.