CAS RN 14681-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-di-tert-Butyl-4-(1-hydroxyethyl)phenol, TRC
CAS: 14681-20-0 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.38 Synonym: 3,5-Bis(1,1-dimethylethyl)-4-hydroxy-alpha-methylbenzenemethanol,2,6-Di-tert-butyl-4-(1-hydroxyethyl)phenol,2,6-Di-tert-butyl-4-(alpha-hydroxyethyl)phenol SMILES: CC(O)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C