missing translation for 'onlineSavingsMsg'
Learn More

17beta-Estradiol Dimethyl Ether, TRC

Product Code. 30008198
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008198 2.5 g 2.5g
30005351 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008198 Supplier Toronto Research Chemicals Supplier No. TRCE8880352.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4954-14-7
Molecular Formula C20 H28 O2
Molecular Weight (g/mol) 300.44
Synonym (17β)-3,17-Dimethoxyestra-1,3,5(10)-triene, Estra-1,3,5(10)-triene, 3,17β-dimethoxy- (6CI,7CI,8CI), 17β-Dimethylestradiol, 3,17β-Dimethoxyestra-1,3,5(10)-triene, Estradiol dimethyl ether, Methylestradiol methyl ether
IUPAC Name (8R,9S,13S,14S,17S)-3,17-dimethoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene
SMILES CO[C@H]1CC[C@H]2[C@@H]3CCc4cc(OC)ccc4[C@H]3CC[C@]12C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C20H28O2/c1-20-11-10-16-15-7-5-14(21-2)12-13(15)4-6-17(16)18(20)8-9-19(20)22-3/h5,7,12,16-19H,4,6,8-11H2,1-3H3/t16-,17-,18+,19+,20+/m1/s1
Formula Weight 300.2089
Percent Purity >95%
Chemical Name or Material 17b-Estradiol Dimethyl Ether
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.