CAS RN 4954-14-7
CAS RN 4954-14-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
17beta-Estradiol Dimethyl Ether, TRC
CAS: 4954-14-7 Molecular Formula: C20 H28 O2 Molecular Weight (g/mol): 300.44 Synonym: (17β)-3,17-Dimethoxyestra-1,3,5(10)-triene,Estra-1,3,5(10)-triene, 3,17β-dimethoxy- (6CI,7CI,8CI),17β-Dimethylestradiol,3,17β-Dimethoxyestra-1,3,5(10)-triene,Estradiol dimethyl ether,Methylestradiol methyl ether IUPAC Name: (8R,9S,13S,14S,17S)-3,17-dimethoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene SMILES: CO[C@H]1CC[C@H]2[C@@H]3CCc4cc(OC)ccc4[C@H]3CC[C@]12C