missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dioxopyrrolidin-1-yl dodecanoate, TRC

Product Code. 30387668
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387668 100 mg -
30376705 500 mg -
30388208 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387668 Supplier Toronto Research Chemicals Supplier No. TRCD495508100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14565-47-0
Molecular Formula C16 H27 N O4
Molecular Weight (g/mol) 297.39
Synonym 2,5-Pyrrolidinedione, 1-[(1-oxododecyl)oxy]- (9CI), Succinimide, N-(lauroyloxy)- (8CI), Lauric acid, O-succinimido deriv. (8CI), N-(Lauroyloxy)succinimide, N-Succinimidyl dodecanoate, N-Succinimidyl laurate, Succinimidyl dodecanoate
IUPAC Name (2,5-dioxopyrrolidin-1-yl) dodecanoate
SMILES CCCCCCCCCCCC(=O)ON1C(=O)CCC1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H27NO4/c1-2-3-4-5-6-7-8-9-10-11-16(20)21-17-14(18)12-13-15(17)19/h2-13H2,1H3
Formula Weight 297.194
Chemical Name or Material 2,5-Dioxopyrrolidin-1-yl dodecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.