CAS RN 14565-47-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,5-Dioxopyrrolidin-1-yl dodecanoate, TRC
CAS: 14565-47-0 Molecular Formula: C16 H27 N O4 Molecular Weight (g/mol): 297.39 Synonym: 2,5-Pyrrolidinedione, 1-[(1-oxododecyl)oxy]- (9CI),Succinimide, N-(lauroyloxy)- (8CI),Lauric acid, O-succinimido deriv. (8CI),N-(Lauroyloxy)succinimide,N-Succinimidyl dodecanoate,N-Succinimidyl laurate,Succinimidyl dodecanoate IUPAC Name: (2,5-dioxopyrrolidin-1-yl) dodecanoate SMILES: CCCCCCCCCCCC(=O)ON1C(=O)CCC1=O