missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dinitrophenol (Wetted with water>15%), TRC

Product Code. 30378146
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378146 500 mg -
30368856 100 mg 100mg
30386633 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378146 Supplier Toronto Research Chemicals Supplier No. TRCD494358500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-71-5
Molecular Formula C6 H4 N2 O5
Molecular Weight (g/mol) 184.11
Synonym Phenol, 2,5-dinitro-, 2,5-Dinitrophenol, 2,5-DNP, NSC 90441, Phenol, γ-dinitro-, γ-Dinitrophenol
IUPAC Name 2,5-dinitrophenol
SMILES Oc1cc(ccc1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H4N2O5/c9-6-3-4(7(10)11)1-2-5(6)8(12)13/h1-3,9H
Formula Weight 184.012
Chemical Name or Material 2,5-Dinitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.