accessibility menu, dialog, popup

UserName

OHNOONOO

CAS RN 329-71-5

IUPAC Name: 2,5-dinitrophenol
Synonyms: 2,5-dinitrophenol
Molecular Weight (g/mol): 184.11
Molecular Formula: C6H4N2O5
InChi Key: UWEZBKLLMKVIPI-UHFFFAOYSA-N
SMILES: OC1=CC(=CC=C1[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)