missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzyl alcohol, 99%

Product Code. 10562781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10562781 25 mL Glass bottle 25mL
10038771 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10562781 Supplier Thermo Scientific Acros Supplier No. 182200250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33524-31-1 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33524-31-1
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004615
InChI Key WGQMUABRZGUAOS-UHFFFAOYSA-N
Synonym 2,5-dimethoxybenzyl alcohol, 2,5-dimethoxyphenyl methanol, benzenemethanol, 2,5-dimethoxy, acmc-1ckmt, 2,5-dimethoxybenzylalcohol, 2,5-dimethoxylbenzylalcohol, benzenemethanol,2,5-dimethoxy, 2,5-dimethoxyphenyl-methanol, wgqmuabrzguaos-uhfffaoysa, 2,5-dimethoxyphenyl methanol #
PubChem CID 118495
IUPAC Name (2,5-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)OC)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 122°C to 125°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2OH
Refractive Index 1.5460 to 1.548
Quantity 25 mL
Formula Weight 168.19
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 2, 5-Dimethoxybenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.