accessibility menu, dialog, popup

UserName

H3COOCH3HO

CAS RN 33524-31-1

IUPAC Name: (2,5-dimethoxyphenyl)methanol
Synonyms: (2,5-dimethoxyphenyl)methanol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: WGQMUABRZGUAOS-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(CO)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)