missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzyl alcohol, 99%

Product Code. 10038771 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10038771

Brand: Thermo Scientific Acros 182200050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33524-31-1 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33524-31-1
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004615
InChI Key WGQMUABRZGUAOS-UHFFFAOYSA-N
Synonym 2,5-dimethoxybenzyl alcohol, 2,5-dimethoxyphenyl methanol, benzenemethanol, 2,5-dimethoxy, acmc-1ckmt, 2,5-dimethoxybenzylalcohol, 2,5-dimethoxylbenzylalcohol, benzenemethanol,2,5-dimethoxy, 2,5-dimethoxyphenyl-methanol, wgqmuabrzguaos-uhfffaoysa, 2,5-dimethoxyphenyl methanol #
PubChem CID 118495
IUPAC Name (2,5-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)OC)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 122°C to 125°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2OH
Refractive Index 1.5460 to 1.548
Quantity 5 mL
Formula Weight 168.19
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 2, 5-Dimethoxybenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.