missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzaldehyde, 97%

Product Code. 15494297
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494297 1 g 1g
15404307 5 g 5g
15414307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15494297 Supplier Thermo Scientific Alfa Aesar Supplier No. H31689.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74553-29-0 | C7H4Br2O | 263.916 g/mol

2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74553-29-0
Molecular Formula C7H4Br2O
Molecular Weight (g/mol) 263.916
MDL Number MFCD00462864
InChI Key BQBXKWGMPUCSQV-UHFFFAOYSA-N
Synonym benzaldehyde, 2,5-dibromo, 2,5-dibromo-benzaldehyde, zlchem 813, acmc-20a0sm, 2,5-di bromobenzaldehyde, ksc493q1h, 2,5-dibromobenzaldehyde
PubChem CID 13540069
IUPAC Name 2,5-dibromobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)Br

Specifications

Melting Point 88°C to 102°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 263.93
Percent Purity 97%
Chemical Name or Material 2,5-Dibromobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.