CAS RN 74553-29-0
IUPAC Name:
2,5-dibromobenzaldehyde
Synonyms:
2,5-dibromobenzaldehyde
Molecular Weight (g/mol):
263.92
Molecular Formula:
C7H4Br2O
InChi Key:
BQBXKWGMPUCSQV-UHFFFAOYSA-N
SMILES:
BrC1=CC(C=O)=C(Br)C=C1
1
–
1
of
1
results
1
2,5-Dibromobenzaldehyde, 97%
CAS: 74553-29-0 Molecular Formula: C7H4Br2O Molecular Weight (g/mol): 263.916 MDL Number: MFCD00462864 InChI Key: BQBXKWGMPUCSQV-UHFFFAOYSA-N Synonym: benzaldehyde, 2,5-dibromo,2,5-dibromo-benzaldehyde,zlchem 813,acmc-20a0sm,2,5-di bromobenzaldehyde,ksc493q1h,2,5-dibromobenzaldehyde PubChem CID: 13540069 IUPAC Name: 2,5-dibromobenzaldehyde SMILES: C1=CC(=C(C=C1Br)C=O)Br