missing translation for 'onlineSavingsMsg'
Learn More

2,4-Octanedione, 98+%, Thermo Scientific™

Product Code. 11449442
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449442 250 g 250g
11429442 10 g 10g
11439442 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449442 Supplier Thermo Scientific Alfa Aesar Supplier No. A10161.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14090-87-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00015259
InChI Key GJYXGIIWJFZCLN-UHFFFAOYSA-N
Synonym 2,4-octanedione, 2,4-octanedion, acmc-1bu2j
PubChem CID 84192
IUPAC Name octane-2,4-dione
SMILES CCCCC(=O)CC(C)=O

Specifications

Density 0.923
Boiling Point 97°C to 99°C (27 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.459
Quantity 250 g
Beilstein 1751147
Formula Weight 142.2
Percent Purity ≥98%
Chemical Name or Material 2,4-Octanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.