accessibility menu, dialog, popup

UserName

CAS RN 14090-87-0

H3COCH3O

CAS RN 14090-87-0

IUPAC Name: octane-2,4-dione
Synonyms: octane-2,4-dione
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: GJYXGIIWJFZCLN-UHFFFAOYSA-N
SMILES: CCCCC(=O)CC(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)