missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15400686
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15400686 5 g 5g
15410686 250 mg 250mg
15490676 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 15400686

Brand: Thermo Scientific Alfa Aesar H54038.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54001-16-0
Molecular Formula C12H11NO3S
Molecular Weight (g/mol) 249.284
MDL Number MFCD06857928
InChI Key HWHHZTHVYFDJNK-UHFFFAOYSA-N
Synonym 2-4-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-4-methoxyphenyl-4-methylthiazole-5-carboxylic acid, 2-4-methoxyphenyl-4-methyl-5-thiazolecarboxylic acid, 4-methyl-2-4-methoxyphenyl thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-4-methoxyphenyl-4-methyl, 2-4-methoxy phenyl-4-methyl-1,3-thiazol-5-carboxylic acid
PubChem CID 4962843
IUPAC Name 2-(4-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)OC)C(=O)O

Specifications

Melting Point 236°C to 241°C
Quantity 5 g
Formula Weight 249.28
Percent Purity 97%
Chemical Name or Material 2-(4-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.