accessibility menu, dialog, popup

UserName

CAS RN 54001-16-0

H3COSOOH3CN

CAS RN 54001-16-0

IUPAC Name: 2-(4-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
Synonyms: 2-(4-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 248.28
Molecular Formula: C12H10NO3S
InChi Key: HWHHZTHVYFDJNK-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1)C1=NC(C)=C(S1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results