missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isopropylphenyl)propanoic acid, TRC

Product Code. 30400579
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400579 10 mg -
30405436 100 mg 100mg
30404402 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400579 Supplier Toronto Research Chemicals Supplier No. TRCI82489510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3585-48-6
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.25
Synonym Benzeneacetic acid, alpha-methyl-4-(1-methylethyl)-, Hydratropic acid, p-isopropyl- (8CI), alpha-Methyl-4-(1-methylethyl)benzeneacetic acid, 2-(p-Isopropylphenyl)propionic acid, 2-(4-Isopropylphenyl)propionic acid, p-Isopropylhydratropic acid
SMILES CC(C)c1ccc(cc1)C(C)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H16O2/c1-8(2)10-4-6-11(7-5-10)9(3)12(13)14/h4-9H,1-3H3,(H,13,14)
Formula Weight 192.115
Chemical Name or Material 2-(4-Isopropylphenyl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.