CAS RN 3585-48-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(4-Isopropylphenyl)propanoic acid, TRC
CAS: 3585-48-6 Molecular Formula: C12H16O2 Molecular Weight (g/mol): 192.25 Synonym: Benzeneacetic acid, alpha-methyl-4-(1-methylethyl)-,Hydratropic acid, p-isopropyl- (8CI),alpha-Methyl-4-(1-methylethyl)benzeneacetic acid,2-(p-Isopropylphenyl)propionic acid,2-(4-Isopropylphenyl)propionic acid,p-Isopropylhydratropic acid SMILES: CC(C)c1ccc(cc1)C(C)C(=O)O