missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzenesulfonyl chloride, 98%

Product Code. 11489450
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489450 5 g 5g
11479450 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489450 Supplier Thermo Scientific Alfa Aesar Supplier No. H50301.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 609-60-9 | C8H9ClO2S | 204.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-60-9
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.67
MDL Number MFCD00156143
InChI Key FREOGXBZEAMJQN-UHFFFAOYSA-N
Synonym 2,4-dimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-benzenesulfonyl chloride, 2,4-dimethylphenyl chlorosulfone, 2,4-dimethylbenzenesulfonylchloride, acmc-1avoq, ksc355s4b, tos-bb-1127, 2,4-dimethylphenylsulfonyl chloride, 2,4-dimethyl benzene sulfonyl chloride
PubChem CID 69109
SMILES CC1=CC=C(C(C)=C1)S(Cl)(=O)=O

Specifications

Melting Point 32°C to 33°C
Quantity 5 g
Odor Acrid
Refractive Index 1.555
UN Number UN3261
IUPAC Name 2,4-dimethylbenzenesulfonyl chloride
Formula Weight 204.68
Percent Purity 98%
Chemical Name or Material 2,4-Dimethylbenzenesulfonyl chloride

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.