accessibility menu, dialog, popup

UserName

CH3H3CSClOO

CAS RN 609-60-9

IUPAC Name: 2,4-dimethylbenzene-1-sulfonyl chloride
Synonyms: 2,4-dimethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: FREOGXBZEAMJQN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(C)=C1)S(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results