missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)ethylboronic acid pinacol ester, 98%

Product Code. 15487755
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487755 5 g 5g
15477755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487755 Supplier Thermo Scientific Alfa Aesar Supplier No. H30303.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 444094-88-6 | C14H20BClO2 | 266.572 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444094-88-6
Molecular Formula C14H20BClO2
Molecular Weight (g/mol) 266.572
MDL Number MFCD12546184
InChI Key BUSKOHHHQUMTIT-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl ethylboronic acid pinacol ester, 2-2-4-chlorophenyl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-chlorophenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-chlorophenylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-2-4-chlorophenyl ethyl-4,4,5,5-tetramethyl
PubChem CID 46221962
IUPAC Name 2-[2-(4-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2=CC=C(C=C2)Cl

Specifications

Melting Point 34°C to 38°C
Quantity 5 g
Formula Weight 266.58
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenyl)ethylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.