accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CCl

CAS RN 444094-88-6

IUPAC Name: 2-[2-(4-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[2-(4-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 266.57
Molecular Formula: C14H20BClO2
InChi Key: BUSKOHHHQUMTIT-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCC2=CC=C(Cl)C=C2)OC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results