missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)ethylboronic acid pinacol ester, 98%

Product Code. 15477755
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15477755 1 g 1g
15487755 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Produktkod. 15477755 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H30303.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

CAS: 444094-88-6 | C14H20BClO2 | 266.572 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 444094-88-6
Molecular Formula C14H20BClO2
Molecular Weight (g/mol) 266.572
MDL Number MFCD12546184
InChI Key BUSKOHHHQUMTIT-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl ethylboronic acid pinacol ester, 2-2-4-chlorophenyl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-chlorophenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-chlorophenylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-2-4-chlorophenyl ethyl-4,4,5,5-tetramethyl
PubChem CID 46221962
IUPAC Name 2-[2-(4-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2=CC=C(C=C2)Cl

Specifikationer

Melting Point 34°C to 38°C
Quantity 1 g
Formula Weight 266.58
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenyl)ethylboronic acid pinacol ester

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.