missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)-6-methoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 11451380
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11451380 5 g 5g
11441380 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11451380

Brand: Thermo Scientific Alfa Aesar H26419.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180134-14-9
Molecular Formula C10H6F6O2
Molecular Weight (g/mol) 272.146
MDL Number MFCD06660316
InChI Key IEENEJLKQKCCDD-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl-6-methoxybenzaldehyde, 2-methoxy-4,6-bis trifluoromethyl benzaldehyde
PubChem CID 17750804
IUPAC Name 2-methoxy-4,6-bis(trifluoromethyl)benzaldehyde
SMILES COC1=CC(=CC(=C1C=O)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 36°C to 39°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 272.15
Percent Purity 97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)-6-methoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.