CAS RN 180134-14-9
CAS RN 180134-14-9
IUPAC Name:
2-methoxy-4,6-bis(trifluoromethyl)benzaldehyde
Synonyms:
2-methoxy-4,6-bis(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
272.15
Molecular Formula:
C10H6F6O2
InChi Key:
IEENEJLKQKCCDD-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC(=C1C=O)C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
2,4-Bis(trifluoromethyl)-6-methoxybenzaldehyde, 97%, Thermo Scientific™
CAS: 180134-14-9 Molecular Formula: C10H6F6O2 Molecular Weight (g/mol): 272.146 MDL Number: MFCD06660316 InChI Key: IEENEJLKQKCCDD-UHFFFAOYSA-N Synonym: 2,4-bis trifluoromethyl-6-methoxybenzaldehyde,2-methoxy-4,6-bis trifluoromethyl benzaldehyde PubChem CID: 17750804 IUPAC Name: 2-methoxy-4,6-bis(trifluoromethyl)benzaldehyde SMILES: COC1=CC(=CC(=C1C=O)C(F)(F)F)C(F)(F)F