missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenylmagnesium bromide, 1M in 2-MeTHF, Thermo Scientific Chemicals

Catalog Number 15490996
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

2,4,6-Trimethylphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
CAS 2633-66-1
Molecular Formula C9H11BrMg
MDL Number MFCD00009663
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 2-mesitylmagnesium bromide, mesitylmagnesium bromide, 2,4,6-trimethylphenylmagnesium bromide, 2-mesitylmagnesium bromide solution, mesmgbr, yxvsitsudrgill-uhfffaoysa-m, magnesium,bromo 2,4,6-trimethylphenyl, 2-mesitylmagnesium bromide solution, 1.0 m in thf, 2,4,6-trimethylphenylmagnesium bromide, 1m in 2-methf, 2-mesitylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
Solubility Information It reacts with water.
InChI Key VZMGGVXGYWKQPB-UHFFFAOYSA-M
SMILES CC1=CC([Mg]Br)=CC(C)=C1C
IUPAC Name magnesium;1,3,5-trimethylbenzene-6-ide;bromide
Molecular Weight (g/mol) 223.40
PubChem CID 5098599
Formula Weight 223.4
Concentration or Composition (by Analyte or Components) 1M in 2-MeTHF
Chemical Name or Material 2,4,6-Trimethylphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'