missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxypyrimidine, 98%, Thermo Scientific™

Product Code. 10051304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10051304 250 mg 250mg
10091254 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10051304 Supplier Thermo Scientific Acros Supplier No. 426742500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13106-85-9
Molecular Formula C7H10N2O3
Molecular Weight (g/mol) 170.17
InChI Key RJVAFLZWVUIBOU-UHFFFAOYSA-N
Synonym acmc-209bkb, 2,4,6-trimethoxy-pyrimidine, pyrimidine,2,4,6-trimethoxy, rjvaflzwvuibou-uhfffaoysa, 2,4,6-trimethoxypyrimidine
PubChem CID 726938
IUPAC Name 2,4,6-trimethoxypyrimidine
SMILES COC1=CC(=NC(=N1)OC)OC

Specifications

Melting Point 51°C to 54°C
Boiling Point 232°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 250 mg
Formula Weight 170.17
Percent Purity 98%
Chemical Name or Material 2, 4, 6-Trimethoxypyrimidine
compliance-icons
Product Identifier
  • 2,4,6-Trimethoxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.