accessibility menu, dialog, popup

UserName

CH3ONNOCH3OCH3

CAS RN 13106-85-9

IUPAC Name: 2,4,6-trimethoxypyrimidine
Synonyms: 2,4,6-trimethoxypyrimidine
Molecular Weight (g/mol): 170.17
Molecular Formula: C7H10N2O3
InChi Key: RJVAFLZWVUIBOU-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(OC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results