missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trihydroxybenzaldehyde, 95%

Product Code. 11494217
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494217 25 g 25g
11474217 1 g 1g
11484217 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494217 Supplier Thermo Scientific Alfa Aesar Supplier No. A19678.14
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 487-70-7 | C7H6O4 | 154.12 g/mol

2,4,6-Trihydroxybenzaldehyde is used in the preparation of 2-methyl-phloroglucinol by hydrogenation using 5% palladium/carbon. It acts as a reactant for the synthesis of biologically active molecules such as 2,4,6-trichlorophenyl hydrazones. It is also used as a xanthine oxidase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trihydroxybenzaldehyde is used in the preparation of 2-methyl-phloroglucinol by hydrogenation using 5% palladium/carbon. It acts as a reactant for the synthesis of biologically active molecules such as 2,4,6-trichlorophenyl hydrazones. It is also used as a xanthine oxidase inhibitor.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-70-7
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00003329
InChI Key BTQAJGSMXCDDAJ-UHFFFAOYSA-N
Synonym phloroglucinaldehyde, benzaldehyde, 2,4,6-trihydroxy, formylphloroglucinal, phloroglucinolcarboxaldehyde, formylphloroglucinol, phloroglucinol aldehyde, 2,4,6-trihydroxy-benzaldehyde, 2,4,6-trihydroxybenzaldehyd, pubchem21790, acmc-1ao01
PubChem CID 68099
IUPAC Name 2,4,6-trihydroxybenzaldehyde
SMILES OC1=CC(O)=C(C=O)C(O)=C1

Specifications

Melting Point ∼195°C (decomposition)
Quantity 25 g
Beilstein 2254429
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 154.12
Percent Purity 95%
Chemical Name or Material 2,4,6-Trihydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.