accessibility menu, dialog, popup

UserName

CAS RN 487-70-7

IUPAC Name: 2,4,6-trihydroxybenzaldehyde
Synonyms: 2,4,6-trihydroxybenzaldehyde
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: BTQAJGSMXCDDAJ-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C(C=O)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results