Learn More
2,4,6-Trichloroaniline, 98+%, Thermo Scientific Chemicals
£25.10 - £166.00
Chemical Identifiers
CAS | 634-93-5 |
---|---|
Molecular Formula | C6H4Cl3N |
Molecular Weight (g/mol) | 196.455 |
MDL Number | MFCD00007663 |
InChI Key | NATVSFWWYVJTAZ-UHFFFAOYSA-N |
Synonym | s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974 |
PubChem CID | 12471 |
IUPAC Name | 2,4,6-trichloroaniline |
SMILES | C1=C(C=C(C(=C1Cl)N)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449346
|
Thermo Scientific Alfa Aesar
A19044.14 |
25 g |
£25.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459346
|
Thermo Scientific Alfa Aesar
A19044.22 |
100 g |
£61.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469346
|
Thermo Scientific Alfa Aesar
A19044.36 |
500 g | |||||||
Description
Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
634-93-5 | |
196.455 | |
NATVSFWWYVJTAZ-UHFFFAOYSA-N | |
12471 | |
C1=C(C=C(C(=C1Cl)N)Cl)Cl |
C6H4Cl3N | |
MFCD00007663 | |
s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974 | |
2,4,6-trichloroaniline |
Specifications
75°C to 79°C | |
262°C | |
MFCD00007663 | |
879568 | |
Insoluble in water. | |
C1=C(C=C(C(=C1Cl)N)Cl)Cl | |
196.455 | |
196.46 | |
2,4,6-Trichloroaniline |
634-93-5 | |
C6H4Cl3N | |
UN2018 | |
s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974 | |
NATVSFWWYVJTAZ-UHFFFAOYSA-N | |
2,4,6-trichloroaniline | |
12471 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID
missing translation for 'einecsNumber' : 211-219-8
missing translation for 'rtecsNumber' : BZ0250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only