missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloroaniline, 98+%

Product Code. 11459346
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459346 100 g 100g
11449346 25 g 25g
11469346 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459346 Supplier Thermo Scientific Alfa Aesar Supplier No. A19044.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 634-93-5 | C6H4Cl3N | 196.455 g/mol

Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-93-5
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
MDL Number MFCD00007663
InChI Key NATVSFWWYVJTAZ-UHFFFAOYSA-N
Synonym s-trichloroaniline, benzenamine, 2,4,6-trichloro, aniline, 2,4,6-trichloro, sym-trichloroaniline, 2,4,6-trichlorobenzenamine, 2,4,6-trichlorophenylamine, 1-amino-2,4,6-trichlorobenzene, unii-j7ic72n9b0, ccris 601, pubchem9974
PubChem CID 12471
IUPAC Name 2,4,6-trichloroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)Cl

Specifications

Melting Point 75°C to 79°C
Quantity 100 g
UN Number UN2018
Beilstein 879568
Boiling Point 262°C
Solubility Information Insoluble in water.
Formula Weight 196.46
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trichloroaniline
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trichloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,6-Trichloroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.