missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxycinnamic acid, predominantly trans, 98+%

Product Code. 11418476
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418476 10 g 10g
11428476 50 g 50g
11438476 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418476 Supplier Thermo Scientific Alfa Aesar Supplier No. A15936.09
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7345-82-6 | C11H12O4 | 208.213 g/mol

2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2E,2E)-N,N,N-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2′E,2′′E)-N,N′,N′′-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7345-82-6
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00004376
InChI Key QAXPUWGAGVERSJ-VOTSOKGWSA-N
Synonym 2,3-dimethoxycinnamic acid, trans-2,3-dimethoxycinnamic acid, 2e-3-2,3-dimethoxyphenyl acrylic acid, e-3-2,3-dimethoxyphenyl acrylic acid, 2,3-dimethoxy cinnamic acid, 3-2,3-dimethoxyphenyl acrylic acid, 2e-3-2,3-dimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3-dimethoxyphenyl, e-3-2,3-dimethoxyphenyl prop-2-enoic acid, e-3-2,3-dimethoxy-phenyl-acrylic acid
PubChem CID 735842
IUPAC Name (E)-3-(2,3-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC(=C1OC)C=CC(=O)O

Specifications

Melting Point 179°C to 183°C
Quantity 10 g
Beilstein 2695122
Formula Weight 208.21
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxycinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.