missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10405702
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10405702 250 mg Amber Glass Bottle 250mg
10702863 1 g Amber Glass Bottle 1g
10716905 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10405702 Supplier Thermo Scientific Maybridge Supplier No. CC23901CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1914-60-9
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD03086168
InChI Key WEVFUSSJCGAVOH-UHFFFAOYNA-N
Synonym 2,3-dihydrobenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 2,3-dihydro, 2,3-dihydro-benzofuran-2-carboxylic acid, 2,3-dihydrobenzo b furan-2-carboxylic acid, 2-benzofurancarboxylicacid, 2,3-dihydro, 2,3-dihydro-2-benzofurancarboxylic acid, dihydrobenzofuran-2-carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylic, 2,3-dihydrobenzofuran carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylicacid
PubChem CID 2776555
IUPAC Name 2,3-dihydro-1-benzofuran-2-carboxylic acid
SMILES OC(=O)C1CC2=CC=CC=C2O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.