accessibility menu, dialog, popup

UserName

OOHO

CAS RN 1914-60-9

IUPAC Name: 2,3-dihydro-1-benzofuran-2-carboxylic acid
Synonyms: 2,3-dihydro-1-benzofuran-2-carboxylic acid
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: WEVFUSSJCGAVOH-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results