missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyridine-4-boronic acid, 95%, Thermo Scientific™

Product Code. 15443747
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15443747

Brand: Thermo Scientific Alfa Aesar H27668.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in HPLC and NMR chromatography. It is involved in several cross-coupling reactions.

Solubility
Soluble in organic solvents.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951677-39-7
Molecular Formula C5H4BCl2NO2
Molecular Weight (g/mol) 191.80
MDL Number MFCD04038756
InChI Key ABRTZUPJNVJMHP-UHFFFAOYSA-N
Synonym 2,3-dichloropyridine-4-boronic acid, 2,3-dichloro-4-pyridineboronic acid, 2,3-dichloropyridin-4-yl boronic acid, 2,3-dichloropyridine-4-boronicacid, 2,3-dichloro-4-pyridineboronicacid, pubchem7941, 2,3-dichloropyridin-4-yl-4-boronic acid, acmc-209rv3, 4-borono-2,3-dichloropyridine, 2,3-dichloro-4-pyridylboronic acid
PubChem CID 3478955
IUPAC Name (2,3-dichloropyridin-4-yl)boronic acid
SMILES OB(O)C1=C(Cl)C(Cl)=NC=C1

Specifications

Melting Point 136°C to 138°C
Quantity 250 mg
Solubility Information Soluble in organic solvents.
Formula Weight 191.81
Percent Purity 95%
Chemical Name or Material 2,3-Dichloropyridine-4-boronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.